Search
Search

| Assay Name: | hsa-miR-219a-1-3p |
|---|---|
| miRBase Accession Number: | MI0000296 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAGUUGAGUCUGGACGUCCCG |
| Species: | Human, Bovine, Dasypus novemcinctus, Pan troglodytes, Pig, Pteropus alecto |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000044, mir-219 |
Mature miRNA Sequence: | AGAGUUGAGUCUGGACGUCCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-219a-1-3p | MIMAT0004567 |
| Bovine | bta-miR-219 | MIMAT0030444 |
| Dasypus novemcinctus | dno-miR-219a-3p | MIMAT0047772 |
| Pan troglodytes | ptr-miR-219-1-3p | MIMAT0008074 |
| Pig | ssc-miR-219a | MIMAT0020590 |
| Pteropus alecto | pal-miR-219a-3p | MIMAT0040071 |