Search
Search

| Assay Name: | hsa-miR-509-3-5p |
|---|---|
| miRBase Accession Number: | MI0005717 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUGCAGACGUGGCAAUCAUG |
| Species: | Human, Gorilla gorilla |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UACUGCAGACGUGGCAAUCAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-509-3-5p | MIMAT0004975 |
| Gorilla gorilla | ggo-miR-509 | MIMAT0024234 |