Search
Search

| Assay Name: | mmu-miR-26b-3p |
|---|---|
| miRBase Accession Number: | MI0000575 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUGUUCUCCAUUACUUGGCUC |
| Species: | Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Rabbit |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000043, mir-26 |
Mature miRNA Sequence: | CCUGUUCUCCAUUACUUGGCUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-26b-3p | MIMAT0004630 |
| Rat | rno-miR-26b-3p | MIMAT0004714 |
| Cricetulus griseus | cgr-miR-26b-3p | MIMAT0023873 |
| Dasypus novemcinctus | dno-miR-26b-3p | MIMAT0047542 |
| Goat | chi-miR-26b-3p | MIMAT0036104 |
| Guinea pig | cpo-miR-26b-3p | MIMAT0046889 |
| View More Rows | ||
| Rabbit | ocu-miR-26b-3p | MIMAT0048138 |