Search
Search

| Assay Name: | hsa-miR-33a-3p |
|---|---|
| miRBase Accession Number: | MI0000091 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAUGUUUCCACAGUGCAUCAC |
| Species: | Human, Mouse |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000070, mir-33 |
Mature miRNA Sequence: | CAAUGUUUCCACAGUGCAUCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-33a-3p | MIMAT0004506 |
| Mouse | mmu-miR-33-3p | MIMAT0004666 |