Search
Search

| Assay Name: | hsa-miR-340-5p |
|---|---|
| miRBase Accession Number: | MI0000802 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAUAAAGCAAUGAGACUGAUU |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000191, mir-340 |
Mature miRNA Sequence: | UUAUAAAGCAAUGAGACUGAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-340-5p | MIMAT0004692 |
| Mouse | mmu-miR-340-5p | MIMAT0004651 |
| Rat | rno-miR-340-5p | MIMAT0004650 |
| Callithrix jacchus | cja-miR-340 | MIMAT0049315 |
| Cricetulus griseus | cgr-miR-340-5p | MIMAT0023920 |
| Dasypus novemcinctus | dno-miR-340-5p | MIMAT0047823 |
| View More Rows | ||
| Daubentonia madagascariensis | dma-miR-340 | MIMAT0049244 |
| Dog | cfa-miR-340 | MIMAT0009892 |
| Goat | chi-miR-340-5p | MIMAT0036155 |
| Guinea pig | cpo-miR-340-5p | MIMAT0047178 |
| Horse | eca-miR-340-5p | MIMAT0013066 |
| Microcebus murinus | mmr-miR-340 | MIMAT0049159 |
| Nomascus leucogenys | nle-miR-340 | MIMAT0049343 |
| Otolemur garnettii | oga-miR-340 | MIMAT0049696 |
| Pan paniscus | ppa-miR-340 | MIMAT0049106 |
| Pan troglodytes | ptr-miR-340 | MIMAT0008109 |
| Papio hamadryas | pha-miR-340 | MIMAT0049556 |
| Pig | ssc-miR-340 | MIMAT0013891 |
| Pongo pygmaeus | ppy-miR-340 | MIMAT0015843 |
| Pteropus alecto | pal-miR-340-5p | MIMAT0040191 |
| Rabbit | ocu-miR-340-5p | MIMAT0048386 |
| Rhesus monkey | mml-miR-340-5p | MIMAT0006281 |