Search
Search

| Assay Name: | hsa-miR-138-2-3p |
|---|---|
| miRBase Accession Number: | MI0000455 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUAUUUCACGACACCAGGGUU |
| Species: | Human, Dasypus novemcinctus, Pteropus alecto |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000075, mir-138 |
Mature miRNA Sequence: | GCUAUUUCACGACACCAGGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-138-2-3p | MIMAT0004596 |
| Dasypus novemcinctus | dno-miR-138-2-3p | MIMAT0047650 |
| Pteropus alecto | pal-miR-138-2-3p | MIMAT0040153 |