Search
Search

| Assay Name: | hsa-miR-31-3p |
|---|---|
| miRBase Accession Number: | MI0000089 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGCUAUGCCAACAUAUUGCCAU |
| Species: | Human, Pteropus alecto |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | UGCUAUGCCAACAUAUUGCCAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-31-3p | MIMAT0004504 |
| Pteropus alecto | pal-miR-31-3p | MIMAT0040137 |