Search
Search

| Assay Name: | rno-miR-673-5p |
|---|---|
| miRBase Accession Number: | MI0006158 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCACAGCUCCGGUCCUUGGAG |
| Species: | Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000405, mir-673 |
Mature miRNA Sequence: | CUCACAGCUCCGGUCCUUGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-673-5p | MIMAT0005328 |