Search
Search

| Assay Name: | rno-miR-224-5p |
|---|---|
| miRBase Accession Number: | MI0003483 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAGUCACUAGUGGUUCCGUUU |
| Species: | Rat, Dog, Goat, Nomascus leucogenys |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000088, mir-224 |
Mature miRNA Sequence: | CAAGUCACUAGUGGUUCCGUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-224-5p | MIMAT0003119 |
| Dog | cfa-miR-224 | MIMAT0006744 |
| Goat | chi-miR-224-5p | MIMAT0036073 |
| Nomascus leucogenys | nle-miR-224 | MIMAT0049400 |