Search
Search

| Assay Name: | hsa-miR-1236-3p |
|---|---|
| miRBase Accession Number: | MI0006326 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUCUUCCCCUUGUCUCUCCAG |
| Species: | Human, Pongo pygmaeus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000689, mir-1236 |
Mature miRNA Sequence: | CCUCUUCCCCUUGUCUCUCCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1236-3p | MIMAT0005591 |
| Pongo pygmaeus | ppy-miR-1236 | MIMAT0016166 |