Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM14692
          Assay Name: hsa-miR-365a-5p
          miRBase Accession Number: MI0000767
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 14309285 - 14309371 [+] on Build GRCh38
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Species: Human, Mouse, Bovine, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Ornithorhynchus anatinus, Rabbit
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM14692
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000061, mir-365

          Mature miRNA Sequence:

          AGGGACUUUUGGGGGCAGAUGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-365a-5p MIMAT0009199
          Mouse mmu-miR-365-1-5p MIMAT0017077
          Bovine bta-miR-365-5p MIMAT0012539
          Columba livia cli-miR-365-5p MIMAT0038624
          Cricetulus griseus cgr-miR-365-5p MIMAT0023935
          Dasypus novemcinctus dno-miR-365-5p MIMAT0047835
          View More Rows
          Guinea pig cpo-miR-365-5p MIMAT0047194
          Ornithorhynchus anatinus oan-miR-365-1-5p MIMAT0006822
          Rabbit ocu-miR-365-5p MIMAT0048399

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-365a
          Stem-loop Accession # MI0000767
          Stem-loop Sequence
          ACCGCAGGGAAAAUGAGGGACUUUUGGGGGCAGAUGUGUUUCCAUUCCACUAUCAUAAUGCCCCUAAAAAUCCUUAUUGCUCUUGCA
          Chromosome Location Chr. 16 - 14309285 - 14309371 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0009199
          miRBase ID: hsa-miR-365a-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: Chr. 16 - 14309285 - 14309371 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-365-1
          Stem-loop Accession # MI0000768
          Stem-loop Sequence
          ACCGCAGGGAAAAUGAGGGACUUUUGGGGGCAGAUGUGUUUCCAUUCCGCUAUCAUAAUGCCCCUAAAAAUCCUUAUUGCUCUUGCA
          Chromosome Location Chr. 16 - 13453840 - 13453926 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017077
          miRBase ID: mmu-miR-365-1-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: Chr. 16 - 13453840 - 13453926 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-365-1
          Stem-loop Accession # MI0005465
          Stem-loop Sequence
          ACCGCAGGGAAAAUGAGGGACUUUUGGGGGCAGAUGUGUUUGCAUUCCACUAUCAUAAUGCCCCUAAAAAUCCUUAUUGCUCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012539
          miRBase ID: bta-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-365-1
          Stem-loop Accession # MI0030035
          Stem-loop Sequence
          ACCGCAGGGAAAAUGAGGGACUUUUGGGGGCAGAUGUGUUUCCAUUACACUAUCAUAAUGCCCCUAAAAAUCCUUAUUGCUCUUGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038624
          miRBase ID: cli-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-365
          Stem-loop Accession # MI0020511
          Stem-loop Sequence
          GGGAAAAUGAGGGACUUUUGGGGGCAGAUGUGUUUCCAUUCCGCUAUCAUAAUGCCCCUAAAAAUCCUUAUUGCUCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023935
          miRBase ID: cgr-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-365-1
          Stem-loop Accession # MI0039082
          Stem-loop Sequence
          AGGGACUUUUGGGGGCAGAUGUGUUUCCAUUCCACUAUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047835
          miRBase ID: dno-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-365-1
          Stem-loop Accession # MI0038745
          Stem-loop Sequence
          AGGGACUUUUGGGGGCAGAUGUGUUUCCAUUCCACUAUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047194
          miRBase ID: cpo-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-365-1
          Stem-loop Accession # MI0006677
          Stem-loop Sequence
          GCAGGGAAAAUGAGGGACUUUUGGGGGCAGAUGUGUUUCCAUUGCACUAUCAUAAUGCCCCUAAAAAUCCUUAUUGCUCUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006822
          miRBase ID: oan-miR-365-1-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-365-1
          Stem-loop Accession # MI0039380
          Stem-loop Sequence
          AGGGACUUUUGGGGGCAGAUGUGUUUGCAUUCCAGUAUCAUAAUGCCCCUAAAAAUCCUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048399
          miRBase ID: ocu-miR-365-5p
          Mature miRNA Sequence: AGGGACUUUUGGGGGCAGAUGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303451_pri
          TaqMan™ Pri-miRNA Assay : Mm03307334_pri
          Ambion® Pre-miR™ miRNA Precursor : PM14692
          TaqMan™ MicroRNA Assay : 007917_mat
          mirVana® miRNA inhibitor : MH14692
          mirVana® miRNA mimic : MC14692
          TaqMan™ Advanced miRNA Assay : 478842_mir
          TaqMan™ Advanced miRNA Assay : mmu482919_mir


          miRBase Alias:

          Mouse: mmu-miR-365-1* (v17)
          Ochrobactrum anthropi: oan-miR-365* (v18)
          Ochrobactrum anthropi: oan-miR-365-5p (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline