Search
Search

| Assay Name: | hsa-miR-365b-5p |
|---|---|
| miRBase Accession Number: | MI0000769 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGACUUUCAGGGGCAGCUGU |
| Species: | Human, Alligator mississippiensis, Columba livia |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000061, mir-365 |
Mature miRNA Sequence: | AGGGACUUUCAGGGGCAGCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-365b-5p | MIMAT0022833 |
| Alligator mississippiensis | ami-miR-365-2-5p | MIMAT0038273 |
| Columba livia | cli-miR-365-2-5p | MIMAT0038626 |