Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM19462
          Assay Name: mmu-miR-224-3p
          miRBase Accession Number: MI0000711
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 72261031 - 72261112 [-] on Build GRCm38
          Mature miRNA Sequence: AAAUGGUGCCCUAGUGACUACA
          Species: Mouse, Rat
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM19462
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000088, mir-224

          Mature miRNA Sequence:

          AAAUGGUGCCCUAGUGACUACA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-224-3p MIMAT0017062
          Rat rno-miR-224-3p MIMAT0017200

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-224
          Stem-loop Accession # MI0000711
          Stem-loop Sequence
          GGGCUUUUAAGUCACUAGUGGUUCCGUUUAGUAGAUGGUUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location Chr. X - 72261031 - 72261112 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017062
          miRBase ID: mmu-miR-224-3p
          Mature miRNA Sequence: AAAUGGUGCCCUAGUGACUACA
          Chromosome Location: Chr. X - 72261031 - 72261112 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-224
          Stem-loop Accession # MI0003483
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGGUUUUUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location Chr. X - 152223754 - 152223835 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017200
          miRBase ID: rno-miR-224-3p
          Mature miRNA Sequence: AAAUGGUGCCCUAGUGACUACA
          Chromosome Location: Chr. X - 152223754 - 152223835 [-] on Build Rnor_6.0


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307192_pri
          TaqMan™ Pri-miRNA Assay : Rn03465521_pri
          Ambion® Pre-miR™ miRNA Precursor : PM19462
          TaqMan™ MicroRNA Assay : 462218_mat
          mirVana® miRNA inhibitor : MH19462
          mirVana® miRNA mimic : MC19462
          TaqMan™ Advanced miRNA Assay : rno481009_mir
          TaqMan™ Advanced miRNA Assay : mmu481009_mir


          miRBase Alias:

          Mouse: mmu-miR-224* (v17)
          Rat: rno-miR-224* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline