Search
Search

| Assay Name: | hsa-miR-548aa |
|---|---|
| miRBase Accession Number: | MI0016689 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAACCACAAUUACUUUUGCACCA |
| Species: | Human |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAACCACAAUUACUUUUGCACCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548aa | MIMAT0018447 |
| Human | hsa-miR-548t-3p | MIMAT0022730 |