Search
Search

| Assay Name: | mmu-miR-128-1-5p |
|---|---|
| miRBase Accession Number: | MI0000155 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGGGCCGUAGCACUGUCUGA |
| Species: | Mouse, Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | CGGGGCCGUAGCACUGUCUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-128-1-5p | MIMAT0016982 |
| Rat | rno-miR-128-1-5p | MIMAT0017118 |