Search
Search

| Assay Name: | mmu-miR-1b-3p |
|---|---|
| miRBase Accession Number: | MI0006283 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGGUACAUAAAGAAGUAUGUGC |
| Species: | Mouse, Cricetulus griseus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000038, mir-1 |
Mature miRNA Sequence: | UGGGUACAUAAAGAAGUAUGUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1b-3p | MIMAT0017326 |
| Cricetulus griseus | cgr-miR-1b | MIMAT0041154 |