Search
Search

| Assay Name: | mmu-miR-181d-3p |
|---|---|
| miRBase Accession Number: | MI0005450 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCCACCGGGGGAUGAAUGUCA |
| Species: | Mouse, Cricetulus griseus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | CCCACCGGGGGAUGAAUGUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-181d-3p | MIMAT0017264 |
| Cricetulus griseus | cgr-miR-181d-3p | MIMAT0023804 |