Search
Search

| Assay Name: | rno-miR-3549 |
|---|---|
| miRBase Accession Number: | MI0015405 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUCUUGCCCGCAAAGACCC |
| Species: | Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000082, mir-193 |
Mature miRNA Sequence: | AUCUUGCCCGCAAAGACCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3549 | MIMAT0017807 |