Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM24181
          Assay Name: hsa-miR-6511b-5p
          miRBase Accession Number: MI0022552
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 2106669 - 2106753 [-] on Build GRCh38
          Mature miRNA Sequence: CUGCAGGCAGAAGUGGGGCUGACA
          Species: Human
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM24181
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0001382, mir-6511

          Mature miRNA Sequence:

          CUGCAGGCAGAAGUGGGGCUGACA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-6511b-5p MIMAT0025847

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-6511b-1
          Stem-loop Accession # MI0022552
          Stem-loop Sequence
          GGGACGGGGCCUGCAGGCAGAAGUGGGGCUGACAGGGCAGAGGGUUGCGCCCCCUCACCACCCCUUCUGCCUGCAGCGGUGGGCU
          Chromosome Location Chr. 16 - 2106669 - 2106753 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0025847
          miRBase ID: hsa-miR-6511b-5p
          Mature miRNA Sequence: CUGCAGGCAGAAGUGGGGCUGACA
          Chromosome Location: Chr. 16 - 15134075 - 15134145 [-] on Build GRCh38

          Stem-loop ID hsa-mir-6511b-2
          Stem-loop Accession # MI0023431
          Stem-loop Sequence
          GGGGCCUGCAGGCAGAAGUGGGGCUGACAGGGCAGAGGGUUGCGCCCCCUCACCACCCCUUCUGCCUGCAG
          Chromosome Location Chr. 16 - 15134075 - 15134145 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0025847
          miRBase ID: hsa-miR-6511b-5p
          Mature miRNA Sequence: CUGCAGGCAGAAGUGGGGCUGACA
          Chromosome Location: Chr. 16 - 15134075 - 15134145 [-] on Build GRCh38


          Back To Top

          More Information




          Related Products

          Ambion® Pre-miR™ miRNA Precursor : PM24181
          TaqMan™ MicroRNA Assay : 473470_mat
          mirVana® miRNA inhibitor : MH24181
          mirVana® miRNA mimic : MC24181
          TaqMan™ Advanced miRNA Assay : 480209_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline