Search
Search

| Assay Name: | hsa-miR-6128 |
|---|---|
| miRBase Accession Number: | MI0021272 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUGGAAUUGGAGUCAAAA |
| Species: | Human, Pan troglodytes, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001442, mir-6128 |
Mature miRNA Sequence: | ACUGGAAUUGGAGUCAAAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6128 | MIMAT0024611 |
| Pan troglodytes | ptr-miR-6128 | MIMAT0024601 |
| Rhesus monkey | mml-miR-6128 | MIMAT0024620 |