Search
Search

| Assay Name: | hsa-let-7b-5p |
|---|---|
| miRBase Accession Number: | MI0000063 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGUAGUAGGUUGUGUGGUU |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Fugu rubripes, Goat, Guinea pig, Metriaclima zebra, Microcebus murinus, Monodelphis domestica, Neolamprologus brichardi, Ophiophagus hannah, Oreochromis niloticus, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Paralichthys olivaceus, Pongo pygmaeus, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Saimiri boliviensis, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGUAGGUUGUGUGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7b-5p | MIMAT0000063 |
| Mouse | mmu-let-7b-5p | MIMAT0000522 |
| Rat | rno-let-7b-5p | MIMAT0000775 |
| Alligator mississippiensis | ami-let-7b-5p | MIMAT0038003 |
| Astatotilapia burtoni | abu-let-7b | MIMAT0042138 |
| Bovine | bta-let-7b | MIMAT0004331 |
| View More Rows | ||
| Callithrix jacchus | cja-let-7b | MIMAT0039325 |
| Chicken | gga-let-7b | MIMAT0001102 |
| Chrysemys picta | cpi-let-7b-5p | MIMAT0037598 |
| Cricetulus griseus | cgr-let-7b | MIMAT0023717 |
| Cyprinus carpio | ccr-let-7b | MIMAT0026190 |
| Dasypus novemcinctus | dno-let-7b-5p | MIMAT0047477 |
| Daubentonia madagascariensis | dma-let-7b | MIMAT0049258 |
| Dog | cfa-let-7b | MIMAT0009836 |
| Fugu rubripes | fru-let-7b | MIMAT0003016 |
| Goat | chi-let-7b-5p | MIMAT0035882 |
| Guinea pig | cpo-let-7b-5p | MIMAT0046820 |
| Metriaclima zebra | mze-let-7b | MIMAT0042376 |
| Microcebus murinus | mmr-let-7b | MIMAT0049187 |
| Monodelphis domestica | mdo-let-7b | MIMAT0004162 |
| Neolamprologus brichardi | nbr-let-7b | MIMAT0042564 |
| Ophiophagus hannah | oha-let-7b-5p | MIMAT0036634 |
| Oreochromis niloticus | oni-let-7b | MIMAT0042743 |
| Ornithorhynchus anatinus | oan-let-7b-5p | MIMAT0006897 |
| Otolemur garnettii | oga-let-7b | MIMAT0049620 |
| Pan paniscus | ppa-let-7b | MIMAT0049092 |
| Pan troglodytes | ptr-let-7b | MIMAT0007937 |
| Papio hamadryas | pha-let-7b | MIMAT0049508 |
| Paralichthys olivaceus | pol-let-7b-5p | MIMAT0025414 |
| Pongo pygmaeus | ppy-let-7b | MIMAT0015722 |
| Pundamilia nyererei | pny-let-7b | MIMAT0042946 |
| Python bivittatus | pbv-let-7b-5p | MIMAT0038796 |
| Rabbit | ocu-let-7b-5p | MIMAT0048078 |
| Rhesus monkey | mml-let-7b-5p | MIMAT0006152 |
| Saimiri boliviensis | sbo-let-7b | MIMAT0049490 |
| Salmo salar | ssa-let-7b-5p | MIMAT0032701 |
| Taeniopygia guttata | tgu-let-7b-5p | MIMAT0014509 |
| Tetraodon nigroviridis | tni-let-7b | MIMAT0003017 |
| Zebrafish | dre-let-7b | MIMAT0001760 |