Search
Search

| Assay Name: | rno-miR-493-3p |
|---|---|
| miRBase Accession Number: | MI0003540 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAAGGUCUACUGUGUGCCAG |
| Species: | Rat, Dog |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000230, mir-493 |
Mature miRNA Sequence: | UGAAGGUCUACUGUGUGCCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-493-3p | MIMAT0003191 |
| Dog | cfa-miR-493 | MIMAT0006711 |