Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11443
          Assay Name: hsa-miR-618
          miRBase Accession Number: MI0003632
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 80935736 - 80935833 [-] on Build GRCh38
          Mature miRNA Sequence: AAACUCUACUUGUCCUUCUGAGU
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11443
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000553, mir-618

          Mature miRNA Sequence:

          AAACUCUACUUGUCCUUCUGAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-618 MIMAT0003287
          Pan troglodytes ptr-miR-618 MIMAT0008280
          Pongo pygmaeus ppy-miR-618 MIMAT0016060
          Rhesus monkey mml-miR-618 MIMAT0006468

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-618
          Stem-loop Accession # MI0003632
          Stem-loop Sequence
          CUCUUGUUCACAGCCAAACUCUACUUGUCCUUCUGAGUGUAAUUACGUACAUGCAGUAGCUCAGGAGACAAGCAGGUUUACCCUGUGGAUGAGUCUGA
          Chromosome Location Chr. 12 - 80935736 - 80935833 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003287
          miRBase ID: hsa-miR-618
          Mature miRNA Sequence: AAACUCUACUUGUCCUUCUGAGU
          Chromosome Location: Chr. 12 - 80935736 - 80935833 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-618
          Stem-loop Accession # MI0008817
          Stem-loop Sequence
          CUCUUGUUCACAGCCAAACUCUACUUGUCCUUCUGAGUGUAAUUACGUACAUGCAGUAGCUCAGGAGACAAGCAGGGUUACCCUGUGGAUGAGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008280
          miRBase ID: ptr-miR-618
          Mature miRNA Sequence: AAACUCUACUUGUCCUUCUGAGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-618
          Stem-loop Accession # MI0015103
          Stem-loop Sequence
          CUCUUGUUCACAGCCAAACUCUACUUGUCCUUCUGAGUGUAAUUAUGUACAUGCAGUAGCUCAGGAGACAAGCAGGGUUACCCUGUGGACGAGUCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016060
          miRBase ID: ppy-miR-618
          Mature miRNA Sequence: AAACUCUACUUGUCCUUCUGAGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-618
          Stem-loop Accession # MI0007871
          Stem-loop Sequence
          UCUUGUUCACAACCAAACUCUACUUGUCCUUCUGAGUGUGAUUACGCCCAUGGAGUAGCUCAGGAGGCAAACAGGGUUACCCUGUGGAUAGGUCUGAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006468
          miRBase ID: mml-miR-618
          Mature miRNA Sequence: AAACUCUACUUGUCCUUCUGAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304649_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11443
          TaqMan™ MicroRNA Assay : 001593
          mirVana® miRNA inhibitor : MH11443
          mirVana® miRNA mimic : MC11443
          TaqMan™ Advanced miRNA Assay : 479101_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline