Search
Search

| Assay Name: | hsa-miR-145-5p |
|---|---|
| miRBase Accession Number: | MI0000461 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUCCAGUUUUCCCAGGAAUCCCU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Daubentonia madagascariensis, Dog, Gadus morhua, Goat, Guinea pig, Horse, Ictalurus punctatus, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Oreochromis niloticus, Otolemur garnettii, Pan paniscus, Papio hamadryas, Petromyzon marinus, Pteropus alecto, Python bivittatus, Rabbit, Salmo salar |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GUCCAGUUUUCCCAGGAAUCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-145-5p | MIMAT0000437 |
| Mouse | mmu-miR-145a-5p | MIMAT0000157 |
| Rat | rno-miR-145-5p | MIMAT0000851 |
| Bovine | bta-miR-145 | MIMAT0003542 |
| Callithrix jacchus | cja-miR-145 | MIMAT0039414 |
| Chrysemys picta | cpi-miR-145-5p | MIMAT0037772 |
| View More Rows | ||
| Columba livia | cli-miR-145-5p | MIMAT0038533 |
| Daubentonia madagascariensis | dma-miR-145 | MIMAT0049256 |
| Dog | cfa-miR-145 | MIMAT0009863 |
| Gadus morhua | gmo-miR-145-5p | MIMAT0044334 |
| Goat | chi-miR-145-5p | MIMAT0035967 |
| Guinea pig | cpo-miR-145-5p | MIMAT0047014 |
| Horse | eca-miR-145 | MIMAT0013064 |
| Ictalurus punctatus | ipu-miR-145 | MIMAT0029422 |
| Microcebus murinus | mmr-miR-145 | MIMAT0049208 |
| Monodelphis domestica | mdo-miR-145-5p | MIMAT0004116 |
| Nomascus leucogenys | nle-miR-145 | MIMAT0049426 |
| Oreochromis niloticus | oni-miR-145 | MIMAT0043395 |
| Otolemur garnettii | oga-miR-145 | MIMAT0049668 |
| Pan paniscus | ppa-miR-145 | MIMAT0049057 |
| Papio hamadryas | pha-miR-145 | MIMAT0049513 |
| Petromyzon marinus | pma-miR-145-5p | MIMAT0019476 |
| Pteropus alecto | pal-miR-145-5p | MIMAT0040158 |
| Python bivittatus | pbv-miR-145-5p | MIMAT0038959 |
| Rabbit | ocu-miR-145-5p | MIMAT0048251 |
| Salmo salar | ssa-miR-145-5p | MIMAT0032369 |