Search
Search

| Assay Name: | mmu-miR-675-5p |
|---|---|
| miRBase Accession Number: | MI0004123 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUGCGGAAAGGGCCCACAGU |
| Species: | Mouse, Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000365, mir-675 |
Mature miRNA Sequence: | UGGUGCGGAAAGGGCCCACAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-675-5p | MIMAT0003725 |
| Rat | rno-miR-675-5p | MIMAT0012841 |