Search
Search

| Assay Name: | hsa-miR-193a-5p |
|---|---|
| miRBase Accession Number: | MI0000487 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGGUCUUUGCGGGCGAGAUGA |
| Species: | Human, Bovine, Chicken, Dasypus novemcinctus, Dog, Guinea pig, Horse, Ophiophagus hannah, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000082, mir-193 |
Mature miRNA Sequence: | UGGGUCUUUGCGGGCGAGAUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-193a-5p | MIMAT0004614 |
| Bovine | bta-miR-193a-5p | MIMAT0003794 |
| Chicken | gga-miR-193a-5p | MIMAT0026787 |
| Dasypus novemcinctus | dno-miR-193a-5p | MIMAT0047721 |
| Dog | cfa-miR-193a | MIMAT0006735 |
| Guinea pig | cpo-miR-193a-5p | MIMAT0047070 |
| View More Rows | ||
| Horse | eca-miR-193a-5p | MIMAT0013025 |
| Ophiophagus hannah | oha-miR-193-5p | MIMAT0036779 |
| Pig | ssc-miR-193a-5p | MIMAT0013894 |
| Pongo pygmaeus | ppy-miR-193a-5p | MIMAT0015787 |
| Pteropus alecto | pal-miR-193a-5p | MIMAT0039931 |
| Python bivittatus | pbv-miR-193a-5p | MIMAT0038989 |
| Rabbit | ocu-miR-193a-5p | MIMAT0048303 |
| Rhesus monkey | mml-miR-193a-5p | MIMAT0006225 |
| Taeniopygia guttata | tgu-miR-193b-5p | MIMAT0014654 |