Search
Search

| Assay Name: | rno-miR-101b-3p |
|---|---|
| miRBase Accession Number: | MI0000648 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACAGUACUGUGAUAGCUGAA |
| Species: | Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000046, mir-101 |
Mature miRNA Sequence: | UACAGUACUGUGAUAGCUGAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-101b-3p | MIMAT0000615 |