Search
Search

| Assay Name: | hsa-miR-221-5p |
|---|---|
| miRBase Accession Number: | MI0000298 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCUGGCAUACAAUGUAGAUUU |
| Species: | Human, Alligator mississippiensis, Columba livia, Paralichthys olivaceus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000051, mir-221 |
Mature miRNA Sequence: | ACCUGGCAUACAAUGUAGAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-221-5p | MIMAT0004568 |
| Alligator mississippiensis | ami-miR-221-5p | MIMAT0038252 |
| Columba livia | cli-miR-221-5p | MIMAT0038604 |
| Paralichthys olivaceus | pol-miR-221-5p | MIMAT0025448 |