Search
Search

| Assay Name: | mmu-miR-301b-3p |
|---|---|
| miRBase Accession Number: | MI0004122 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUGCAAUGGUAUUGUCAAAGC |
| Species: | Mouse, Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | CAGUGCAAUGGUAUUGUCAAAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-301b-3p | MIMAT0004186 |
| Rat | rno-miR-301b-3p | MIMAT0005304 |