Search
Search

| Assay Name: | mmu-miR-196a-2-3p |
|---|---|
| miRBase Accession Number: | MI0000553 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGGCAACAAGAAACUGCCUGA |
| Species: | Mouse, Rat, Cricetulus griseus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UCGGCAACAAGAAACUGCCUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-196a-2-3p | MIMAT0004618 |
| Rat | rno-miR-196a-3p | MIMAT0004737 |
| Cricetulus griseus | cgr-miR-196a-3p | MIMAT0023832 |