Search
Search

| Assay Name: | hsa-miR-340-3p |
|---|---|
| miRBase Accession Number: | MI0000802 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCGUCUCAGUUACUUUAUAGC |
| Species: | Human, Mouse, Goat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000191, mir-340 |
Mature miRNA Sequence: | UCCGUCUCAGUUACUUUAUAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-340-3p | MIMAT0000750 |
| Mouse | mmu-miR-340-3p | MIMAT0000586 |
| Goat | chi-miR-340-3p | MIMAT0036156 |