Search
Search

| Assay Name: | hsa-miR-1260a |
|---|---|
| miRBase Accession Number: | MI0006394 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUCCCACCUCUGCCACCA |
| Species: | Human, Pongo pygmaeus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0001020, mir-1260a |
Mature miRNA Sequence: | AUCCCACCUCUGCCACCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1260a | MIMAT0005911 |
| Pongo pygmaeus | ppy-miR-1260a | MIMAT0016184 |