Search
Search

| Assay Name: | hsa-miR-1267 |
|---|---|
| miRBase Accession Number: | MI0006404 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUGUUGAAGUGUAAUCCCCA |
| Species: | Human, Pan troglodytes |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000640, mir-1267 |
Mature miRNA Sequence: | CCUGUUGAAGUGUAAUCCCCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1267 | MIMAT0005921 |
| Pan troglodytes | ptr-miR-1267 | MIMAT0007991 |