Search
Search

| Assay Name: | hsa-miR-1301-3p |
|---|---|
| miRBase Accession Number: | MI0003815 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAGCUGCCUGGGAGUGACUUC |
| Species: | Human, Guinea pig, Pongo pygmaeus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000742, mir-1301 |
Mature miRNA Sequence: | UUGCAGCUGCCUGGGAGUGACUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1301-3p | MIMAT0005797 |
| Guinea pig | cpo-miR-1301-3p | MIMAT0047420 |
| Pongo pygmaeus | ppy-miR-1301 | MIMAT0016221 |