Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM14755
          Assay Name: mmu-miR-223-5p
          miRBase Accession Number: MI0000703
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 96242817 - 96242926 [+] on Build GRCm38
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Species: Mouse, Rat, Dasypus novemcinctus, Guinea pig, Ornithorhynchus anatinus, Rabbit
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM14755
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000067, mir-223

          Mature miRNA Sequence:

          CGUGUAUUUGACAAGCUGAGUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-223-5p MIMAT0017056
          Rat rno-miR-223-5p MIMAT0017165
          Dasypus novemcinctus dno-miR-223-5p MIMAT0047779
          Guinea pig cpo-miR-223-5p MIMAT0047134
          Ornithorhynchus anatinus oan-miR-223-5p MIMAT0007019
          Rabbit ocu-miR-223-5p MIMAT0048358

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-223
          Stem-loop Accession # MI0000703
          Stem-loop Sequence
          UCUGGCCAUCUGCAGUGUCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCUGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGUGGCUCAUGCCUAUCAG
          Chromosome Location Chr. X - 96242817 - 96242926 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017056
          miRBase ID: mmu-miR-223-5p
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Chromosome Location: Chr. X - 96242817 - 96242926 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-223
          Stem-loop Accession # MI0000963
          Stem-loop Sequence
          UCUGGCCUUCUGCAGUGUUACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCUGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGUGGCUCAUGCUUAUCAG
          Chromosome Location Chr. X - 65367812 - 65367921 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017165
          miRBase ID: rno-miR-223-5p
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Chromosome Location: Chr. X - 65367812 - 65367921 [+] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-223
          Stem-loop Accession # MI0039054
          Stem-loop Sequence
          CGUGUAUUUGACAAGCUGAGUUGGACACUCCAUAUGGUAGAGUGUCAGUUUGUCAAAUACCCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047779
          miRBase ID: dno-miR-223-5p
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-223
          Stem-loop Accession # MI0038715
          Stem-loop Sequence
          CGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047134
          miRBase ID: cpo-miR-223-5p
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-223
          Stem-loop Accession # MI0006796
          Stem-loop Sequence
          GCGACCACGUGACCACACCGCAGUGCCGCGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCGUGUGGUUUGAGUGUCAGUUUGUCAAAUACCCUAAGUGAGGCAUGCUCACCCAUGUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007019
          miRBase ID: oan-miR-223-5p
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-223
          Stem-loop Accession # MI0039359
          Stem-loop Sequence
          CGUGUAUUUGACAAGCUGAGUUGGACACUCCGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048358
          miRBase ID: ocu-miR-223-5p
          Mature miRNA Sequence: CGUGUAUUUGACAAGCUGAGUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465479_pri
          TaqMan™ Pri-miRNA Assay : Mm03307156_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM14755
          TaqMan™ MicroRNA Assay : 007896_mat
          mirVana® miRNA inhibitor : MH14755
          mirVana® miRNA mimic : MC14755
          TaqMan™ Advanced miRNA Assay : rno481008_mir
          TaqMan™ Advanced miRNA Assay : mmu481008_mir


          miRBase Alias:

          Mouse: mmu-miR-223* (v17)
          Rat: rno-miR-223* (v18)
          Ochrobactrum anthropi: oan-miR-223* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0