Search
Search

| Assay Name: | rno-miR-128-2-5p |
|---|---|
| miRBase Accession Number: | MI0000901 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGCCGAUGCACUGUAAGA |
| Species: | Rat, Cricetulus griseus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | GGGGGCCGAUGCACUGUAAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-128-2-5p | MIMAT0017119 |
| Cricetulus griseus | cgr-miR-128 | MIMAT0041168 |