Search
Search

| Assay Name: | rno-miR-673-3p |
|---|---|
| miRBase Accession Number: | MI0006158 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCGGGACUGAGUUCUGUGCAC |
| Species: | Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000405, mir-673 |
Mature miRNA Sequence: | UCCGGGACUGAGUUCUGUGCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-673-3p | MIMAT0017313 |