Search
Search

| Assay Name: | mmu-miR-326-5p |
|---|---|
| miRBase Accession Number: | MI0000598 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGCAGGGCCUUUGUGAAGGCG |
| Species: | Mouse, Guinea pig, Rabbit |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000143, mir-326 |
Mature miRNA Sequence: | GGGGGCAGGGCCUUUGUGAAGGCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-326-5p | MIMAT0017027 |
| Guinea pig | cpo-miR-326-5p | MIMAT0047160 |
| Rabbit | ocu-miR-326-5p | MIMAT0048374 |