Search
Search

| Assay Name: | hsa-miR-6889-3p |
|---|---|
| miRBase Accession Number: | MI0022736 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUGUGCCCCUACUUCCCAG |
| Species: | Human |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UCUGUGCCCCUACUUCCCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6889-3p | MIMAT0027679 |