Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@3ac8d09c
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Order Status
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
PIK3R1
Entrez Gene ID
5295
Gene Name:
phosphoinositide-3-kinase regulatory subunit 1
Gene Aliases
AGM7, GRB1, IMD36, p85, p85-ALPHA
Gene Chromosome Location
Chr.5: on Build GRCh38
UniGene
Hs.132225
Species:
Human
Primer Chromosome Location
Chr.5: 68279524-68280037 on Build GRCh38
View in UCSC browser
Amplicon Length
514
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00260853_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
PIK3R1
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_181523.2
NP_852664.1
7342613
c.562C>T
p.R188C
6932327
c.547G>A
p.A183T
7342612
c.574G>A
p.D192N
3429594
c.623C>A
p.S208Y
389861
c.540C>T
p.H180H
6746568
c.532G>A
p.D178N
6573142
c.613G>A
p.E205K
7346056
c.608delA
p.Y203fs*4
6980936
c.627A>T
p.L209F
1581456
c.629C>A
p.A210D
5019326
c.621T>C
p.I207I
6196522
c.578_579insT
p.L193fs*13
1581455
c.514G>A
p.V172M
241138
c.545delT
p.L182fs*25
6171584
c.603C>T
p.A201A
5633724
c.526A>G
p.M176V
5794093
c.589G>A
p.V197I
6171583
c.610A>T
p.S204C
3071849
c.537G>T
p.V179V
6971613
c.608A>G
p.Y203C
6933741
c.505A>G
p.T169A
6966391
c.503A>G
p.D168G
3975610
c.553G>A
p.A185T
5946061
c.580C>A
p.P194T
XM_005248542.3
XP_005248599.1
-
-
-
XM_011543493.2
XP_011541795.1
-
-
-
XM_017009585.1
XP_016865074.1
-
-
-
XM_017009586.1
XP_016865075.1
-
-
-
GenBank
AK299743.1
-
-
-
BC094795.1
AAH94795.1
-
-
-
ENST00000521381.0
-
-
-
M61906.1
-
-
-
Phenotype
Literature Links:
PIK3R1
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
enzyme modulator
kinase modulator
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-green:8080/10.253.226.236:8080.
git-commit: cebdaefe8eed8bf9cdce87ec1e9ab74c86511abf
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/1.47.0-2022.08.32-1.0