Assay Details
Gene Information
MAPK10 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001318067.1 | NP_001304996.1 | - | - | - |
NM_001318068.1 | NP_001304997.1 | - | - | - | |
NM_001318069.1 | NP_001304998.1 | - | - | - | |
NM_002753.4 | NP_002744.1 | - | - | - | |
NM_138980.3 | NP_620446.1 | - | - | - | |
NM_138982.3 | NP_620448.1 | - | - | - | |
XM_005263129.2 | XP_005263186.1 | - | - | - | |
XM_005263130.2 | XP_005263187.1 | - | - | - | |
XM_005263131.2 | XP_005263188.1 | - | - | - | |
XM_005263135.3 | XP_005263192.1 | - | - | - | |
XM_006714268.2 | XP_006714331.1 | - | - | - | |
XM_006714269.2 | XP_006714332.1 | - | - | - | |
XM_011532117.2 | XP_011530419.1 | - | - | - | |
XM_011532118.2 | XP_011530420.1 | - | - | - | |
XM_011532120.2 | XP_011530422.1 | - | - | - | |
XM_011532121.2 | XP_011530423.1 | - | - | - | |
XM_017008419.1 | XP_016863908.1 | - | - | - | |
XM_017008420.1 | XP_016863909.1 | - | - | - | |
XM_017008421.1 | XP_016863910.1 | - | - | - | |
XM_017008422.1 | XP_016863911.1 | - | - | - | |
XM_017008423.1 | XP_016863912.1 | - | - | - | |
XM_017008424.1 | XP_016863913.1 | - | - | - | |
XM_017008425.1 | XP_016863914.1 | - | - | - | |
XM_017008426.1 | XP_016863915.1 | - | - | - | |
XM_017008427.1 | XP_016863916.1 | - | - | - | |
XM_017008428.1 | XP_016863917.1 | - | - | - | |
XM_017008429.1 | XP_016863918.1 | - | - | - | |
XM_017008430.1 | XP_016863919.1 | - | - | - | |
XM_017008431.1 | XP_016863920.1 | - | - | - | |
XM_017008432.1 | XP_016863921.1 | - | - | - | |
XM_017008433.1 | XP_016863922.1 | - | - | - | |
XM_017008434.1 | XP_016863923.1 | - | - | - | |
XM_017008435.1 | XP_016863924.1 | - | - | - | |
XM_017008436.1 | XP_016863925.1 | - | - | - | |
XM_017008437.1 | XP_016863926.1 | - | - | - | |
XM_017008438.1 | XP_016863927.1 | - | - | - | |
XM_017008439.1 | XP_016863928.1 | - | - | - | |
XM_017008440.1 | XP_016863929.1 | - | - | - | |
XM_017008441.1 | XP_016863930.1 | - | - | - | |
XM_017008442.1 | XP_016863931.1 | - | - | - | |
XM_017008443.1 | XP_016863932.1 | - | - | - | |
XM_017008444.1 | XP_016863933.1 | - | - | - | |
XM_017008445.1 | XP_016863934.1 | - | - | - | |
XM_017008446.1 | XP_016863935.1 | - | - | - | |
XM_017008447.1 | XP_016863936.1 | - | - | - | |
XM_017008448.1 | XP_016863937.1 | - | - | - | |
XM_017008449.1 | XP_016863938.1 | - | - | - | |
XM_017008450.1 | XP_016863939.1 | - | - | - | |
XM_017008451.1 | XP_016863940.1 | - | - | - | |
XM_017008452.1 | XP_016863941.1 | - | - | - |
Phenotype
Literature Links: |
MAPK10 PubMed Links |
SNP ID

More Information
Set Membership: |
![]() |
Panther Classification:
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
