Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
USP9Y
Entrez Gene ID
8287
Gene Name:
ubiquitin specific peptidase 9, Y-linked
Gene Aliases
DFFRY, SPGFY2
Gene Chromosome Location
Chr.Y: on Build GRCh38
UniGene
Hs.598540
Species:
Human
Primer Chromosome Location
Chr.Y: 12860338-12860861 on Build GRCh38
View in UCSC browser
Amplicon Length
524
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00403350_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
USP9Y
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_004654.3
NP_004645.2
-
-
-
XM_017030078.1
XP_016885567.1
-
-
-
GenBank
AK057605.1
-
-
-
BI754571.1
-
-
-
CK823170.1
-
-
-
ENST00000338981.0
-
-
-
Phenotype
MIM: 400005
Literature Links:
USP9Y
PubMed Links
SNP ID
rs966125005
rs375206568
rs1017660431
rs199904650
rs576666864
rs767441144
rs985329588
rs953892854
rs567478645
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860686-12860686 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860742-12860742 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global
T (0.01)
(0.99)
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS
T (0.00)
(1.00)
African American - Not Available
YRI (Yoruba) - Not Available
SAS
T (0.00)
(1.00)
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR
T (0.01)
(0.99)
Japanese - Not Available
JPT (Japanese) - Not Available
EUR
T (0.01)
(0.99)
AMR
T (0.06)
(0.94)
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860655-12860655 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860788-12860788 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860454-12860454 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860413-12860413 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global
C (0.00)
(1.00)
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS
C (0.00)
(1.00)
African American - Not Available
YRI (Yoruba) - Not Available
SAS
C (0.00)
(1.00)
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR
C (0.01)
(0.99)
Japanese - Not Available
JPT (Japanese) - Not Available
EUR
C (0.00)
(1.00)
AMR
C (0.00)
(1.00)
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860504-12860504 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860489-12860489 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.Y: 12860732-12860732 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
ubiquitin-dependent protein catabolic process
transforming growth factor beta receptor signaling pathway
spermatogenesis
cell migration
protein deubiquitination
BMP signaling pathway
thiol-dependent ubiquitin-specific protease activity
cysteine-type peptidase activity
co-SMAD binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0