Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EPHA3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_005233.5 | NP_005224.2 | 74528 | c.644C>T | p.T215M |
5447582 | c.642C>A | p.D214E | |||
3427937 | c.573C>G | p.A191A | |||
5898199 | c.499G>A | p.E167K | |||
5608398 | c.597C>T | p.F199F | |||
82125 | c.561T>G | p.G187G | |||
6962957 | c.632T>A | p.M211K | |||
4120478 | c.563C>T | p.A188V | |||
387339 | c.545C>T | p.A182V | |||
7128552 | c.516T>A | p.G172G | |||
6920727 | c.621G>C | p.K207N | |||
6946976 | c.533G>A | p.G178E | |||
302142 | c.634T>A | p.F212I | |||
6975185 | c.633G>T | p.M211I | |||
4150356 | c.500A>G | p.E167G | |||
122861 | c.528G>C | p.K176N | |||
6952139 | c.553G>T | p.D185Y | |||
1670365 | c.514G>T | p.G172C | |||
6654963 | c.522C>T | p.V174V | |||
1251274 | c.485T>G | p.L162R | |||
6925359 | c.598A>T | p.K200* | |||
1251281 | c.620A>C | p.K207T | |||
77653 | c.485T>A | p.L162Q | |||
3427939 | c.600A>C | p.K200N | |||
4120480 | c.620A>T | p.K207M | |||
6956005 | c.571G>T | p.A191S | |||
5530541 | c.625C>T | p.L209L | |||
6928891 | c.637C>T | p.P213S | |||
48321 | c.497C>A | p.T166N | |||
6937500 | c.550C>T | p.Q184* | |||
6011107 | c.640G>A | p.D214N | |||
6958498 | c.577G>A | p.V193M | |||
396050 | c.544G>T | p.A182S | |||
6960328 | c.478C>A | p.R160S | |||
6952141 | c.557T>C | p.V186A | |||
6914251 | c.532G>A | p.G178R | |||
268222 | c.494A>G | p.N165S | |||
280826 | c.621G>A | p.K207K | |||
94195 | c.616G>T | p.V206L | |||
4992057 | c.610T>C | p.F204L | |||
2828291 | c.501G>T | p.E167D | |||
48322 | c.559G>C | p.G187R | |||
6982264 | c.608C>A | p.P203Q | |||
6965736 | c.539_540insT | p.L181fs*47 | |||
5819627 | c.498T>C | p.T166T | |||
1048853 | c.623A>C | p.N208T | |||
6911947 | c.596T>A | p.F199Y | |||
6933069 | c.620A>G | p.K207R | |||
3427935 | c.484C>T | p.L162L | |||
6962905 | c.512T>C | p.V171A | |||
382885 | c.602A>C | p.K201T | |||
6945801 | c.550C>G | p.Q184E | |||
7118197 | c.501G>A | p.E167E | |||
6947164 | c.479G>A | p.R160H | |||
6972853 | c.621G>T | p.K207N | |||
2828295 | c.594C>G | p.Y198* | |||
7316103 | c.560G>T | p.G187V | |||
4498325 | c.517C>T | p.P173S | |||
1633350 | c.631A>G | p.M211V | |||
6923597 | c.641A>G | p.D214G | |||
6941877 | c.603G>T | p.K201N | |||
NM_182644.2 | NP_872585.1 | 7316104 | c.560G>T | p.G187V | |
302143 | c.634T>A | p.F212I | |||
268223 | c.494A>G | p.N165S | |||
6952140 | c.553G>T | p.D185Y | |||
6937501 | c.550C>T | p.Q184* | |||
6923598 | c.641A>G | p.D214G | |||
7128553 | c.516T>A | p.G172G | |||
3427938 | c.573C>G | p.A191A | |||
4498326 | c.517C>T | p.P173S | |||
6945802 | c.550C>G | p.Q184E | |||
6960329 | c.478C>A | p.R160S | |||
6914252 | c.532G>A | p.G178R | |||
4150357 | c.500A>G | p.E167G | |||
6952142 | c.557T>C | p.V186A | |||
387340 | c.545C>T | p.A182V | |||
3427940 | c.600A>C | p.K200N | |||
6925360 | c.598A>T | p.K200* | |||
6941878 | c.603G>T | p.K201N | |||
5608399 | c.597C>T | p.F199F | |||
6928892 | c.637C>T | p.P213S | |||
6975186 | c.633G>T | p.M211I | |||
1251282 | c.620A>C | p.K207T | |||
1670366 | c.514G>T | p.G172C | |||
6962906 | c.512T>C | p.V171A | |||
6947165 | c.479G>A | p.R160H | |||
6982265 | c.608C>A | p.P203Q | |||
6962958 | c.632T>A | p.M211K | |||
731479 | c.608C>T | p.P203L | |||
6933070 | c.620A>G | p.K207R | |||
4120479 | c.563C>T | p.A188V | |||
6965737 | c.539_540insT | p.L181fs*47 | |||
5898200 | c.499G>A | p.E167K | |||
5447583 | c.642C>A | p.D214E | |||
5530542 | c.625C>T | p.L209L | |||
382886 | c.602A>C | p.K201T | |||
6011108 | c.640G>A | p.D214N | |||
2828292 | c.501G>T | p.E167D | |||
1633351 | c.631A>G | p.M211V | |||
6958499 | c.577G>A | p.V193M | |||
5819628 | c.498T>C | p.T166T | |||
4120481 | c.620A>T | p.K207M | |||
6920728 | c.621G>C | p.K207N | |||
280827 | c.621G>A | p.K207K | |||
1048854 | c.623A>C | p.N208T | |||
7118198 | c.501G>A | p.E167E | |||
6956006 | c.571G>T | p.A191S | |||
3427936 | c.484C>T | p.L162L | |||
6972854 | c.621G>T | p.K207N | |||
6654964 | c.522C>T | p.V174V | |||
2828296 | c.594C>G | p.Y198* | |||
1251275 | c.485T>G | p.L162R | |||
4992058 | c.610T>C | p.F204L | |||
396051 | c.544G>T | p.A182S | |||
6946977 | c.533G>A | p.G178E | |||
6911948 | c.596T>A | p.F199Y | |||
XM_005264715.2 | XP_005264772.1 | - | - | - | |
XM_005264716.2 | XP_005264773.1 | - | - | - | |
GenBank | AF213459.1 | AAG43576.1 | - | - | - |
AF213460.1 | AAG43577.1 | - | - | - | |
AK291411.1 | - | - | - | ||
BC026247.1 | - | - | - | ||
BC063282.1 | AAH63282.1 | - | - | - | |
ENST00000336596.0 | - | - | - | ||
HM437236.1 | - | - | - | ||
M83941.1 | AAA58633.1 | - | - | - |
Literature Links: |
EPHA3 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |