Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CELF1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001025596.2 | NP_001020767.1 | 429068 | c.351G>C | p.K117N |
4033203 | c.414G>A | p.R138R | |||
1638852 | c.373C>T | p.R125* | |||
927449 | c.382T>C | p.F128L | |||
40003 | c.347C>T | p.S116F | |||
3979683 | c.412C>T | p.R138W | |||
5094294 | c.389C>T | p.S130L | |||
6626124 | c.311-1G>T | p.? | |||
3719242 | c.311-6T>C | p.? | |||
4886346 | c.343A>G | p.I115V | |||
6068674 | c.370A>G | p.I124V | |||
4671406 | c.374G>A | p.R125Q | |||
6331679 | c.421C>T | p.R141W | |||
NM_001172639.1 | NP_001166110.1 | 6626123 | c.392-4G>T | p.? | |
6068673 | c.448A>G | p.I150V | |||
6626126 | c.467C>T | p.S156L | |||
6331678 | c.499C>T | p.R167W | |||
6626090 | c.490C>T | p.R164W | |||
5831771 | c.429G>C | p.K143N | |||
6626113 | c.452G>A | p.R151Q | |||
NM_001172640.1 | NP_001166111.1 | - | - | - | |
NM_006560.3 | NP_006551.1 | - | - | - | |
NM_198700.2 | NP_941989.1 | 6626125 | c.311-1G>T | p.? | |
3719243 | c.311-6T>C | p.? | |||
6068675 | c.370A>G | p.I124V | |||
4813385 | c.351G>C | p.K117N | |||
3731092 | c.382T>C | p.F128L | |||
6331680 | c.421C>T | p.R141W | |||
3979684 | c.412C>T | p.R138W | |||
4886347 | c.343A>G | p.I115V | |||
4033204 | c.414G>A | p.R138R | |||
5094295 | c.389C>T | p.S130L | |||
4671407 | c.374G>A | p.R125Q | |||
XM_011519848.2 | XP_011518150.1 | - | - | - | |
XM_011519850.2 | XP_011518152.2 | - | - | - | |
XM_011519852.2 | XP_011518154.1 | - | - | - | |
XM_011519854.1 | XP_011518156.1 | - | - | - | |
XM_011519855.1 | XP_011518157.1 | - | - | - | |
XM_011519856.1 | XP_011518158.1 | - | - | - | |
XM_011519857.1 | XP_011518159.1 | - | - | - | |
XM_011519858.1 | XP_011518160.1 | - | - | - | |
XM_011519859.1 | XP_011518161.1 | - | - | - | |
XM_017017101.1 | XP_016872590.1 | - | - | - | |
XM_017017102.1 | XP_016872591.1 | - | - | - | |
XM_017017103.1 | XP_016872592.1 | - | - | - | |
XM_017017104.1 | XP_016872593.1 | - | - | - | |
XM_017017105.1 | XP_016872594.1 | - | - | - | |
XM_017017106.1 | XP_016872595.1 | - | - | - | |
XM_017017107.1 | XP_016872596.1 | - | - | - | |
XM_017017108.1 | XP_016872597.1 | - | - | - | |
XM_017017109.1 | XP_016872598.1 | - | - | - | |
XM_017017110.1 | XP_016872599.1 | - | - | - | |
XM_017017111.1 | XP_016872600.1 | - | - | - | |
XM_017017112.1 | XP_016872601.1 | - | - | - | |
XM_017017113.1 | XP_016872602.1 | - | - | - | |
XM_017017114.1 | XP_016872603.1 | - | - | - | |
XM_017017115.1 | XP_016872604.1 | - | - | - | |
XM_017017116.1 | XP_016872605.1 | - | - | - | |
XM_017017117.1 | XP_016872606.1 | - | - | - | |
XM_017017118.1 | XP_016872607.1 | - | - | - | |
XM_017017119.1 | XP_016872608.1 | - | - | - | |
XM_017017120.1 | XP_016872609.1 | - | - | - | |
XM_017017121.1 | XP_016872610.1 | - | - | - | |
XM_017017122.1 | XP_016872611.1 | - | - | - | |
XM_017017123.1 | XP_016872612.1 | - | - | - | |
XM_017017124.1 | XP_016872613.1 | - | - | - | |
XM_017017125.1 | XP_016872614.1 | - | - | - | |
XM_017017126.1 | XP_016872615.1 | - | - | - | |
XM_017017127.1 | XP_016872616.1 | - | - | - | |
XM_017017128.1 | XP_016872617.1 | - | - | - | |
XM_017017129.1 | XP_016872618.1 | - | - | - | |
XM_017017130.1 | XP_016872619.1 | - | - | - | |
XM_017017131.1 | XP_016872620.1 | - | - | - | |
XM_017017132.1 | XP_016872621.1 | - | - | - | |
XM_017017133.1 | XP_016872622.1 | - | - | - | |
XM_017017134.1 | XP_016872623.1 | - | - | - | |
XM_017017135.1 | XP_016872624.1 | - | - | - | |
GenBank | AB210019.1 | - | - | - | |
AF248648.1 | AAF86230.1 | - | - | - | |
AF267533.1 | AAF78955.1 | - | - | - | |
AF267534.1 | AAF78956.1 | - | - | - | |
AJ007988.1 | CAC20566.1 | - | - | - | |
AK304430.1 | - | - | - | ||
BC031079.1 | AAH31079.1 | - | - | - | |
BC036782.1 | - | - | - | ||
CB960956.1 | - | - | - | ||
DA320861.1 | - | - | - | ||
DB162286.1 | - | - | - | ||
ENST00000358597.0 | - | - | - | ||
U63289.1 | AAC50895.1 | - | - | - |
Literature Links: |
CELF1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |