Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CELF1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001025596.2 | NP_001020767.1 | 6425098 | c.1190G>T | p.G397V |
927445 | c.1281C>T | p.V427V | |||
6518929 | c.1286C>T | p.S429F | |||
6626093 | c.1212C>A | p.F404L | |||
3809339 | c.1243C>T | p.Q415* | |||
3809337 | c.1248C>T | p.D416D | |||
5012324 | c.1250T>C | p.L417P | |||
1354090 | c.1227C>T | p.P409P | |||
7160250 | c.1199G>A | p.G400E | |||
NM_001172639.1 | NP_001166110.1 | 7160249 | c.1277G>A | p.G426E | |
6425097 | c.1268G>T | p.G423V | |||
6518928 | c.1364C>T | p.S455F | |||
5826970 | c.1305C>T | p.P435P | |||
6626092 | c.1290C>A | p.F430L | |||
5831767 | c.1326C>T | p.D442D | |||
5831768 | c.1321C>T | p.Q441* | |||
NM_001172640.1 | NP_001166111.1 | - | - | - | |
NM_006560.3 | NP_006551.1 | - | - | - | |
NM_198700.2 | NP_941989.1 | 4787236 | c.1218C>T | p.P406P | |
7160251 | c.1190G>A | p.G397E | |||
3809338 | c.1239C>T | p.D413D | |||
3809340 | c.1234C>T | p.Q412* | |||
6425099 | c.1181G>T | p.G394V | |||
6518930 | c.1277C>T | p.S426F | |||
5012325 | c.1241T>C | p.L414P | |||
4192003 | c.1272C>T | p.V424V | |||
6626094 | c.1203C>A | p.F401L | |||
XM_011519848.2 | XP_011518150.1 | - | - | - | |
XM_011519850.2 | XP_011518152.2 | - | - | - | |
XM_011519852.2 | XP_011518154.1 | - | - | - | |
XM_011519854.1 | XP_011518156.1 | - | - | - | |
XM_011519855.1 | XP_011518157.1 | - | - | - | |
XM_011519856.1 | XP_011518158.1 | - | - | - | |
XM_011519857.1 | XP_011518159.1 | - | - | - | |
XM_011519858.1 | XP_011518160.1 | - | - | - | |
XM_011519859.1 | XP_011518161.1 | - | - | - | |
XM_017017101.1 | XP_016872590.1 | - | - | - | |
XM_017017102.1 | XP_016872591.1 | - | - | - | |
XM_017017103.1 | XP_016872592.1 | - | - | - | |
XM_017017104.1 | XP_016872593.1 | - | - | - | |
XM_017017105.1 | XP_016872594.1 | - | - | - | |
XM_017017106.1 | XP_016872595.1 | - | - | - | |
XM_017017107.1 | XP_016872596.1 | - | - | - | |
XM_017017108.1 | XP_016872597.1 | - | - | - | |
XM_017017109.1 | XP_016872598.1 | - | - | - | |
XM_017017110.1 | XP_016872599.1 | - | - | - | |
XM_017017111.1 | XP_016872600.1 | - | - | - | |
XM_017017112.1 | XP_016872601.1 | - | - | - | |
XM_017017113.1 | XP_016872602.1 | - | - | - | |
XM_017017114.1 | XP_016872603.1 | - | - | - | |
XM_017017115.1 | XP_016872604.1 | - | - | - | |
XM_017017116.1 | XP_016872605.1 | - | - | - | |
XM_017017117.1 | XP_016872606.1 | - | - | - | |
XM_017017118.1 | XP_016872607.1 | - | - | - | |
XM_017017119.1 | XP_016872608.1 | - | - | - | |
XM_017017120.1 | XP_016872609.1 | - | - | - | |
XM_017017121.1 | XP_016872610.1 | - | - | - | |
XM_017017122.1 | XP_016872611.1 | - | - | - | |
XM_017017123.1 | XP_016872612.1 | - | - | - | |
XM_017017124.1 | XP_016872613.1 | - | - | - | |
XM_017017125.1 | XP_016872614.1 | - | - | - | |
XM_017017126.1 | XP_016872615.1 | - | - | - | |
XM_017017127.1 | XP_016872616.1 | - | - | - | |
XM_017017128.1 | XP_016872617.1 | - | - | - | |
XM_017017129.1 | XP_016872618.1 | - | - | - | |
XM_017017130.1 | XP_016872619.1 | - | - | - | |
XM_017017131.1 | XP_016872620.1 | - | - | - | |
XM_017017132.1 | XP_016872621.1 | - | - | - | |
XM_017017133.1 | XP_016872622.1 | - | - | - | |
XM_017017134.1 | XP_016872623.1 | - | - | - | |
XM_017017135.1 | XP_016872624.1 | - | - | - | |
GenBank | AB210019.1 | - | - | - | |
AF248648.1 | AAF86230.1 | - | - | - | |
AF267533.1 | AAF78955.1 | - | - | - | |
AF267534.1 | AAF78956.1 | - | - | - | |
AJ007988.1 | CAC20566.1 | - | - | - | |
AK304430.1 | - | - | - | ||
AK307679.1 | - | - | - | ||
BC031079.1 | AAH31079.1 | - | - | - | |
BC036782.1 | - | - | - | ||
ENST00000358597.0 | - | - | - | ||
U63289.1 | AAC50895.1 | - | - | - |
Literature Links: |
CELF1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |