Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FYN | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_002037.5 | NP_002028.1 | 4817216 | c.1540G>A | p.E514K |
3857631 | c.1451G>T | p.R484M | |||
6957394 | c.1441C>T | p.R481* | |||
1697763 | c.1532C>T | p.P511L | |||
3697454 | c.1488T>C | p.H496H | |||
1672684 | c.1561G>A | p.E521K | |||
7428649 | c.1440_1453del14 | p.E480fs*13 | |||
5709803 | c.1591T>G | p.Y531D | |||
4911506 | c.1605A>C | p.E535D | |||
5687075 | c.1612T>C | p.*538Q | |||
6871576 | c.1481C>T | p.S494F | |||
483284 | c.1535C>A | p.T512N | |||
6668087 | c.1589A>G | p.Q530R | |||
6172135 | c.1509G>T | p.W503C | |||
5737730 | c.1515G>T | p.K505N | |||
6976976 | c.1582G>C | p.E528Q | |||
4172394 | c.1591T>C | p.Y531H | |||
5709807 | c.1570_1574delTTTAC | p.F524fs*10 | |||
4586156 | c.1483C>T | p.L495L | |||
5907411 | c.1512A>T | p.K504N | |||
1311477 | c.1479C>T | p.I493I | |||
6955359 | c.1514A>G | p.K505R | |||
4776928 | c.1442G>A | p.R481Q | |||
4685937 | c.1514delA | p.K505fs*>33 | |||
5709805 | c.1588C>T | p.Q530* | |||
4160132 | c.1606A>C | p.N536H | |||
6451031 | c.1417C>T | p.R473W | |||
5709801 | c.1594C>T | p.Q532* | |||
6963832 | c.1577C>T | p.A526V | |||
6172133 | c.1420G>T | p.E474* | |||
6668089 | c.1418G>A | p.R473Q | |||
6982061 | c.1420delG | p.E474fs*26 | |||
6934917 | c.1463C>T | p.P488L | |||
4950023 | c.1432C>T | p.Q478* | |||
3156545 | c.1544A>G | p.Y515C | |||
5709809 | c.1566C>G | p.D522E | |||
5019630 | c.1518C>G | p.D506E | |||
357642 | c.1503C>G | p.H501Q | |||
6949655 | c.1514A>T | p.K505M | |||
5009581 | c.1412A>C | p.N471T | |||
6983754 | c.1525G>A | p.E509K | |||
5008894 | c.1452G>A | p.R484R | |||
NM_153047.3 | NP_694592.1 | - | - | - | |
NM_153048.3 | NP_694593.1 | - | - | - | |
XM_005266890.3 | XP_005266947.1 | 6451032 | c.1408C>T | p.R470W | |
4776929 | c.1433G>A | p.R478Q | |||
6172134 | c.1411G>T | p.E471* | |||
4685938 | c.1505delA | p.K502fs*>33 | |||
6976977 | c.1573G>C | p.E525Q | |||
4586157 | c.1474C>T | p.L492L | |||
5709810 | c.1557C>G | p.D519E | |||
6955360 | c.1505A>G | p.K502R | |||
4950024 | c.1423C>T | p.Q475* | |||
5709806 | c.1579C>T | p.Q527* | |||
4911507 | c.1596A>C | p.E532D | |||
5709802 | c.1585C>T | p.Q529* | |||
3857632 | c.1442G>T | p.R481M | |||
5687076 | c.1603T>C | p.*535Q | |||
4817217 | c.1531G>A | p.E511K | |||
5009582 | c.1403A>C | p.N468T | |||
7428650 | c.1431_1444del14 | p.E477fs*13 | |||
4160133 | c.1597A>C | p.N533H | |||
3156546 | c.1535A>G | p.Y512C | |||
6871577 | c.1472C>T | p.S491F | |||
1137416 | c.1526C>A | p.T509N | |||
6668090 | c.1409G>A | p.R470Q | |||
5019631 | c.1509C>G | p.D503E | |||
3697455 | c.1479T>C | p.H493H | |||
6172136 | c.1500G>T | p.W500C | |||
6982062 | c.1411delG | p.E471fs*26 | |||
1697764 | c.1523C>T | p.P508L | |||
6957395 | c.1432C>T | p.R478* | |||
1672685 | c.1552G>A | p.E518K | |||
5709808 | c.1561_1565delTTTAC | p.F521fs*10 | |||
5709804 | c.1582T>G | p.Y528D | |||
6983755 | c.1516G>A | p.E506K | |||
6949656 | c.1505A>T | p.K502M | |||
5008895 | c.1443G>A | p.R481R | |||
4172395 | c.1582T>C | p.Y528H | |||
6963833 | c.1568C>T | p.A523V | |||
6668088 | c.1580A>G | p.Q527R | |||
5737731 | c.1506G>T | p.K502N | |||
6934918 | c.1454C>T | p.P485L | |||
1311478 | c.1470C>T | p.I490I | |||
5907412 | c.1503A>T | p.K501N | |||
XM_005266892.3 | XP_005266949.1 | - | - | - | |
XM_017010650.1 | XP_016866139.1 | - | - | - | |
XM_017010651.1 | XP_016866140.1 | - | - | - | |
XM_017010652.1 | XP_016866141.1 | - | - | - | |
XM_017010653.1 | XP_016866142.1 | - | - | - | |
XM_017010654.1 | XP_016866143.1 | - | - | - | |
GenBank | AB451293.1 | - | - | - | |
AB451426.1 | - | - | - | ||
AI187118.1 | - | - | - | ||
AK056699.1 | - | - | - | ||
BC032496.1 | AAH32496.1 | - | - | - | |
ENST00000354650.0 | - | - | - | ||
M14333.1 | AAC08285.1 | - | - | - | |
M14676.1 | AAA36615.1 | - | - | - | |
S74774.1 | AAB33113.2 | - | - | - |
Literature Links: |
FYN PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |