Assay Details
Gene Information
MAPK10 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001318067.1 | NP_001304996.1 | - | - | - |
NM_001318068.1 | NP_001304997.1 | 1058463 | c.386A>G | p.K129R | |
3669313 | c.495C>A | p.P165P | |||
735433 | c.482G>T | p.G161V | |||
5677397 | c.416C>T | p.P139L | |||
1058461 | c.429G>T | p.K143N | |||
3669311 | c.501C>T | p.L167L | |||
4794415 | c.489C>T | p.T163T | |||
3606773 | c.516C>T | p.L172L | |||
7138713 | c.410C>T | p.P137L | |||
4789519 | c.507A>T | p.P169P | |||
4398707 | c.512C>T | p.S171F | |||
6707745 | c.464A>G | p.N155S | |||
2956684 | c.518T>A | p.F173Y | |||
4561890 | c.467G>A | p.R156Q | |||
1694639 | c.418G>A | p.E140K | |||
6707739 | c.429G>A | p.K143K | |||
4165053 | c.550+5G>A | p.? | |||
282660 | c.534G>T | p.E178D | |||
1619183 | c.368-2A>G | p.? | |||
4126498 | c.458T>C | p.V153A | |||
381169 | c.479C>T | p.A160V | |||
1431532 | c.524C>T | p.A175V | |||
6306392 | c.486C>A | p.L162L | |||
NM_001318069.1 | NP_001304998.1 | - | - | - | |
NM_002753.4 | NP_002744.1 | - | - | - | |
NM_138980.3 | NP_620446.1 | - | - | - | |
NM_138982.3 | NP_620448.1 | - | - | - | |
XM_005263129.2 | XP_005263186.1 | - | - | - | |
XM_005263130.2 | XP_005263187.1 | - | - | - | |
XM_005263131.2 | XP_005263188.1 | - | - | - | |
XM_005263135.3 | XP_005263192.1 | - | - | - | |
XM_006714268.2 | XP_006714331.1 | - | - | - | |
XM_006714269.2 | XP_006714332.1 | - | - | - | |
XM_011532117.2 | XP_011530419.1 | - | - | - | |
XM_011532118.2 | XP_011530420.1 | - | - | - | |
XM_011532120.2 | XP_011530422.1 | - | - | - | |
XM_011532121.2 | XP_011530423.1 | - | - | - | |
XM_017008419.1 | XP_016863908.1 | - | - | - | |
XM_017008420.1 | XP_016863909.1 | - | - | - | |
XM_017008421.1 | XP_016863910.1 | - | - | - | |
XM_017008422.1 | XP_016863911.1 | - | - | - | |
XM_017008423.1 | XP_016863912.1 | - | - | - | |
XM_017008424.1 | XP_016863913.1 | - | - | - | |
XM_017008425.1 | XP_016863914.1 | - | - | - | |
XM_017008426.1 | XP_016863915.1 | - | - | - | |
XM_017008427.1 | XP_016863916.1 | - | - | - | |
XM_017008428.1 | XP_016863917.1 | - | - | - | |
XM_017008429.1 | XP_016863918.1 | - | - | - | |
XM_017008430.1 | XP_016863919.1 | - | - | - | |
XM_017008431.1 | XP_016863920.1 | - | - | - | |
XM_017008432.1 | XP_016863921.1 | - | - | - | |
XM_017008433.1 | XP_016863922.1 | - | - | - | |
XM_017008434.1 | XP_016863923.1 | 1694638 | c.853G>A | p.E285K | |
4794414 | c.924C>T | p.T308T | |||
381168 | c.914C>T | p.A305V | |||
3669310 | c.936C>T | p.L312L | |||
1619182 | c.803-2A>G | p.? | |||
3669312 | c.930C>A | p.P310P | |||
6306391 | c.921C>A | p.L307L | |||
4789518 | c.942A>T | p.P314P | |||
4561889 | c.902G>A | p.R301Q | |||
7138712 | c.845C>T | p.P282L | |||
3606772 | c.951C>T | p.L317L | |||
1431531 | c.959C>T | p.A320V | |||
4165052 | c.985+5G>A | p.? | |||
4398706 | c.947C>T | p.S316F | |||
4126497 | c.893T>C | p.V298A | |||
5677396 | c.851C>T | p.P284L | |||
6707744 | c.899A>G | p.N300S | |||
1058460 | c.864G>T | p.K288N | |||
1058462 | c.821A>G | p.K274R | |||
735432 | c.917G>T | p.G306V | |||
282661 | c.969G>T | p.E323D | |||
2956683 | c.953T>A | p.F318Y | |||
6707738 | c.864G>A | p.K288K | |||
XM_017008435.1 | XP_016863924.1 | - | - | - | |
XM_017008436.1 | XP_016863925.1 | - | - | - | |
XM_017008437.1 | XP_016863926.1 | - | - | - | |
XM_017008438.1 | XP_016863927.1 | - | - | - | |
XM_017008439.1 | XP_016863928.1 | - | - | - | |
XM_017008440.1 | XP_016863929.1 | - | - | - | |
XM_017008441.1 | XP_016863930.1 | - | - | - | |
XM_017008442.1 | XP_016863931.1 | - | - | - | |
XM_017008443.1 | XP_016863932.1 | - | - | - | |
XM_017008444.1 | XP_016863933.1 | - | - | - | |
XM_017008445.1 | XP_016863934.1 | - | - | - | |
XM_017008446.1 | XP_016863935.1 | - | - | - | |
XM_017008447.1 | XP_016863936.1 | - | - | - | |
XM_017008448.1 | XP_016863937.1 | - | - | - | |
XM_017008449.1 | XP_016863938.1 | - | - | - | |
XM_017008450.1 | XP_016863939.1 | - | - | - | |
XM_017008451.1 | XP_016863940.1 | - | - | - | |
XM_017008452.1 | XP_016863941.1 | - | - | - | |
GenBank | AK057723.1 | - | - | - | |
AK091104.1 | - | - | - | ||
AK124791.1 | - | - | - | ||
AK225974.1 | - | - | - | ||
AK294125.1 | - | - | - | ||
BC022492.1 | - | - | - | ||
BC035057.1 | AAH35057.1 | - | - | - | |
BC051731.1 | AAH51731.1 | - | - | - | |
BC065516.1 | AAH65516.1 | - | - | - | |
U07620.1 | AAC50101.1 | - | - | - | |
U34819.1 | AAC50604.1 | - | - | - | |
U34820.1 | AAC50605.1 | - | - | - |
Phenotype
Literature Links: |
MAPK10 PubMed Links |
SNP ID

More Information
Set Membership: |
![]() |
Panther Classification:
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
