Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DLGAP1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001003809.2 | NP_001003809.1 | - | - | - |
NM_001242761.1 | NP_001229690.1 | 6041920 | c.2577T>C | p.P859P | |
6644831 | c.2595C>T | p.P865P | |||
5614561 | c.2674C>T | p.H892Y | |||
6082872 | c.2614G>T | p.G872W | |||
6148644 | c.2604G>T | p.Q868H | |||
6243977 | c.2688C>A | p.A896A | |||
6281725 | c.2612C>T | p.A871V | |||
4483251 | c.2688C>T | p.A896A | |||
3422169 | c.2644G>T | p.E882* | |||
5623619 | c.2783C>G | p.S928C | |||
6082867 | c.2576C>G | p.P859R | |||
4860698 | c.2584C>T | p.H862Y | |||
4863707 | c.2663T>G | p.F888C | |||
6308415 | c.2613G>A | p.A871A | |||
NM_001242762.1 | NP_001229691.1 | 6082870 | c.1628C>G | p.P543R | |
6148647 | c.1656G>T | p.Q552H | |||
6243980 | c.1740C>A | p.A580A | |||
6281728 | c.1664C>T | p.A555V | |||
5614564 | c.1726C>T | p.H576Y | |||
1646892 | c.1636C>T | p.H546Y | |||
4863709 | c.1715T>G | p.F572C | |||
4483254 | c.1740C>T | p.A580A | |||
6644834 | c.1647C>T | p.P549P | |||
3422171 | c.1696G>T | p.E566* | |||
6082875 | c.1666G>T | p.G556W | |||
6308418 | c.1665G>A | p.A555A | |||
1680194 | c.1677C>G | p.D559E | |||
6041923 | c.1629T>C | p.P543P | |||
NM_001242763.1 | NP_001229692.1 | - | - | - | |
NM_001242764.1 | NP_001229693.1 | - | - | - | |
NM_001242765.1 | NP_001229694.1 | - | - | - | |
NM_001242766.1 | NP_001229695.1 | 6308416 | c.1737G>A | p.A579A | |
6082868 | c.1700C>G | p.P567R | |||
1680193 | c.1749C>G | p.D583E | |||
6148645 | c.1728G>T | p.Q576H | |||
6243978 | c.1812C>A | p.A604A | |||
6281726 | c.1736C>T | p.A579V | |||
6644832 | c.1719C>T | p.P573P | |||
3422170 | c.1768G>T | p.E590* | |||
6041921 | c.1701T>C | p.P567P | |||
1147972 | c.1708C>T | p.H570Y | |||
1589440 | c.1787T>G | p.F596C | |||
6082873 | c.1738G>T | p.G580W | |||
4483252 | c.1812C>T | p.A604A | |||
5614562 | c.1798C>T | p.H600Y | |||
NM_001308390.1 | NP_001295319.1 | - | - | - | |
NM_004746.3 | NP_004737.2 | 4483250 | c.2688C>T | p.A896A | |
6644830 | c.2595C>T | p.P865P | |||
5614560 | c.2674C>T | p.H892Y | |||
6082871 | c.2614G>T | p.G872W | |||
1680192 | c.2625C>G | p.D875E | |||
6308414 | c.2613G>A | p.A871A | |||
3422168 | c.2644G>T | p.E882* | |||
709577 | c.2584C>T | p.H862Y | |||
223919 | c.2605G>A | p.D869N | |||
6082866 | c.2576C>G | p.P859R | |||
6148643 | c.2604G>T | p.Q868H | |||
988298 | c.2663T>G | p.F888C | |||
6243976 | c.2688C>A | p.A896A | |||
127812 | c.2640C>A | p.S880S | |||
6281724 | c.2612C>T | p.A871V | |||
269027 | c.2594C>A | p.P865H | |||
6041919 | c.2577T>C | p.P859P | |||
XM_005258171.2 | XP_005258228.1 | - | - | - | |
XM_005258172.1 | XP_005258229.1 | - | - | - | |
XM_005258173.3 | XP_005258230.2 | - | - | - | |
XM_005258174.4 | XP_005258231.1 | - | - | - | |
XM_006722367.3 | XP_006722430.1 | - | - | - | |
XM_017026080.1 | XP_016881569.1 | - | - | - | |
XM_017026081.1 | XP_016881570.1 | - | - | - | |
XM_017026082.1 | XP_016881571.1 | - | - | - | |
XM_017026083.1 | XP_016881572.1 | - | - | - | |
XM_017026084.1 | XP_016881573.1 | - | - | - | |
XM_017026085.1 | XP_016881574.1 | - | - | - | |
GenBank | AB000276.1 | BAA23257.1 | - | - | - |
AB000277.1 | BAA23258.1 | - | - | - | |
AK294717.1 | - | - | - | ||
AK294747.1 | - | - | - | ||
AK294777.1 | - | - | - | ||
AK299880.1 | - | - | - | ||
AK304101.1 | - | - | - | ||
AK316099.1 | - | - | - | ||
BC136453.1 | - | - | - | ||
BC136454.1 | - | - | - | ||
ENST00000315677.0 | - | - | - | ||
U67988.1 | AAC51119.1 | - | - | - |
Literature Links: |
DLGAP1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |