Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277188.1 | NP_001264117.1 | 6, 7 | 1064 | ||
GenBank mRNA | AK039466.1 | - | 7 | 928 | ||
AK039728.1 | - | 6, 7 | 874 | |||
AK078364.1 | - | 6, 7 | 828 | |||
BC065114.1 | - | 6 | 1021 |