siRNA Details
Gene Symbol: Sox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009237.2 | NP_033263.2 | 1 | 626 | ||
GenBank mRNA | AB108674.1 | - | 1 | 401 | ||
BC052024.1 | - | 1 | 623 | |||
BC063061.1 | - | 1 | 620 | |||
BC096018.1 | - | 1 | 584 |

More Information
Other Species Targeted

Panther Classification:
